Product Details

SNP ID
rs59626664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56243191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAATGGCGCCGGGCCGCTCCTGG[C/G]TGTAGGGGCAGGAGGCCCCATGGGG
Phenotype
MIM: 608730
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD52 PubMed Links

Gene Details

Gene
ANKRD52
Gene Name
ankyrin repeat domain 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173595.3 3272 Missense Mutation ACC,AGC T1061S NP_775866.2
XM_011538197.2 3272 Missense Mutation ACC,AGC T1000S XP_011536499.1
XM_017019183.1 3272 Missense Mutation ACC,AGC T1060S XP_016874672.1
XM_017019184.1 3272 Missense Mutation CAC,CAG H1030Q XP_016874673.1
Gene
SLC39A5
Gene Name
solute carrier family 39 member 5
There are no transcripts associated with this gene.

View Full Product Details