Product Details

SNP ID
rs61754169
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56319972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTATGTGTTTTCCGCTGCTTCCG[C/T]GCCAGCGAGGCTTCAGCCAGCAAGA
Phenotype
MIM: 605861
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CNPY2 PubMed Links

Gene Details

Gene
CNPY2
Gene Name
canopy FGF signaling regulator 2
There are no transcripts associated with this gene.

Gene
PAN2
Gene Name
PAN2 poly(A) specific ribonuclease subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127460.2 3227 Silent Mutation GCA,GCG A950A NP_001120932.1
NM_001166279.1 3227 Silent Mutation GCA,GCG A949A NP_001159751.1
NM_014871.4 3227 Silent Mutation GCA,GCG A946A NP_055686.3

View Full Product Details