Product Details

SNP ID
rs60586224
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57611899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCGGGGGGGCCCGGGCGCCATGC[A/G]GGGGGGGCACAAAGGGGGTCGCTGT
Phenotype
MIM: 610215 MIM: 613142
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ARHGEF25 PubMed Links

Gene Details

Gene
ARHGEF25
Gene Name
Rho guanine nucleotide exchange factor 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111270.2 465 Intron NP_001104740.1
NM_182947.3 465 Missense Mutation CAG,CGG Q2R NP_891992.2
Gene
DTX3
Gene Name
deltex E3 ubiquitin ligase 3
There are no transcripts associated with this gene.

Gene
LOC101927583
Gene Name
uncharacterized LOC101927583
There are no transcripts associated with this gene.

Gene
PIP4K2C
Gene Name
phosphatidylinositol-5-phosphate 4-kinase type 2 gamma
There are no transcripts associated with this gene.

Gene
SLC26A10
Gene Name
solute carrier family 26 member 10
There are no transcripts associated with this gene.

View Full Product Details