Product Details

SNP ID
rs60553748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:68845403 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATTATAAAATGAGCTAACAAAC[C/G]AAAGGCAAAATAAAACCGTAAAGCA
Phenotype
MIM: 114860 MIM: 164785
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CPM PubMed Links

Gene Details

Gene
CPM
Gene Name
carboxypeptidase M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005502.2 7282 Intron NP_001005502.1
NM_001874.4 7282 Intron NP_001865.1
NM_198320.3 7282 Intron NP_938079.1
Gene
MDM2
Gene Name
MDM2 proto-oncogene
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145337.2 7282 UTR 3 NP_001138809.1
NM_001145339.2 7282 UTR 3 NP_001138811.1
NM_001145340.2 7282 UTR 3 NP_001138812.1
NM_001278462.1 7282 UTR 3 NP_001265391.1
NM_002392.5 7282 UTR 3 NP_002383.2
XM_005268872.4 7282 UTR 3 XP_005268929.1
XM_006719399.3 7282 UTR 3 XP_006719462.1
XM_006719400.3 7282 UTR 3 XP_006719463.1

View Full Product Details