Product Details

SNP ID
rs57550068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:94334656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCTTTATTTTGGGGTTTGAAGAT[C/T]CAACAGGAGGATTGAATGTCATTTC
Phenotype
MIM: 610769 MIM: 608414
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOC3L PubMed Links

Gene Details

Gene
NOC3L
Gene Name
NOC3 like DNA replication regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022451.10 2443 Missense Mutation GAA,GGA E751G NP_071896.8
XM_017016544.1 2443 Missense Mutation GAA,GGA E536G XP_016872033.1
Gene
PLCE1
Gene Name
phospholipase C epsilon 1
There are no transcripts associated with this gene.

View Full Product Details