Product Details

SNP ID
rs61203890
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:116058904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGAGGCAGATTAAAATCTCAGC[A/G]GACTAACTGGCGAATTTCCCACAAA
Phenotype
MIM: 601496
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GFRA1 PubMed Links

Gene Details

Gene
GFRA1
Gene Name
GDNF family receptor alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145453.2 7231 UTR 3 NP_001138925.1
NM_005264.5 7231 UTR 3 NP_005255.1
NM_145793.3 7231 UTR 3 NP_665736.1
XM_005269696.1 7231 UTR 3 XP_005269753.1
XM_011539634.1 7231 UTR 3 XP_011537936.1

View Full Product Details