Product Details

SNP ID
rs56772609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:119502680 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCAGGACTCGACCCCGAGATCC[C/T]GAGATCCCAAGGCTGCCAGCAAGCC
Phenotype
MIM: 602856
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RGS10 PubMed Links

Gene Details

Gene
RGS10
Gene Name
regulator of G-protein signaling 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005339.1 Intron NP_001005339.1
NM_002925.3 Intron NP_002916.1
XM_017016492.1 Intron XP_016871981.1
XM_017016493.1 Intron XP_016871982.1

View Full Product Details