Product Details

SNP ID
rs55725412
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:119652620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCTTACAGTTGGCCATTGCCTT[A/G]CCACATTGCCTCACACTGGAGAGAA
Phenotype
MIM: 603883
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
BAG3 PubMed Links

Gene Details

Gene
BAG3
Gene Name
BCL2 associated athanogene 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004281.3 Intron NP_004272.2
XM_005270287.1 Intron XP_005270344.1

View Full Product Details