Product Details

SNP ID
rs62216220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:41799257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTAGATTCTGCAAAGGCAAAAA[G/T]AAATCGATACTGATTTTTAAAAATA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRDM15 PubMed Links

Gene Details

Gene
PRDM15
Gene Name
PR domain 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040424.2 5691 UTR 3 NP_001035514.1
NM_001282934.1 5691 UTR 3 NP_001269863.1
NM_022115.4 5691 Intron NP_071398.3
XM_006724039.2 5691 UTR 3 XP_006724102.1
XM_006724040.2 5691 UTR 3 XP_006724103.1
XM_011529674.1 5691 UTR 3 XP_011527976.1
XM_011529675.1 5691 UTR 3 XP_011527977.1
XM_011529676.2 5691 UTR 3 XP_011527978.1
XM_011529677.2 5691 UTR 3 XP_011527979.1
XM_011529678.2 5691 UTR 3 XP_011527980.1
XM_011529679.2 5691 UTR 3 XP_011527981.1
XM_011529681.2 5691 UTR 3 XP_011527983.1
XM_011529683.1 5691 UTR 3 XP_011527985.1
XM_017028425.1 5691 UTR 3 XP_016883914.1
XM_017028426.1 5691 UTR 3 XP_016883915.1

View Full Product Details