Product Details

SNP ID
rs59811201
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:19945626 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGTACAGGATCTATGGATTGAA[C/T]AGACTTTTTTTCATGGAACTACACA
Phenotype
MIM: 609708
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LPL PubMed Links

Gene Details

Gene
LPL
Gene Name
lipoprotein lipase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000237.2 Intron NP_000228.1

View Full Product Details