Product Details

SNP ID
rs56190382
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:40481610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGACTTGCAGCAGGTCCTCCTCA[C/T]TAAGGAGGCTTCAGGCTGCTCTTGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZFP69 PubMed Links

Gene Details

Gene
ZFP69
Gene Name
ZFP69 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320178.1 Intron NP_001307107.1
NM_001320179.1 Intron NP_001307108.1
NM_198494.2 Intron NP_940896.2
XM_005270809.4 Intron XP_005270866.1
XM_006710605.3 Intron XP_006710668.1
XM_006710606.3 Intron XP_006710669.1

View Full Product Details