Product Details

SNP ID
rs56303620
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:45804039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGGAGGCAGCGGCTGGAGGAGCG[G/T]ACGGGCCCCGCGGGGCCCGAGGGCA
Phenotype
MIM: 612257
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MAST2 PubMed Links

Gene Details

Gene
MAST2
Gene Name
microtubule associated serine/threonine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319245.1 427 Silent Mutation CGG,CGT R48R NP_001306174.1
NM_001324320.1 427 Silent Mutation CGG,CGT R48R NP_001311249.1
NM_001324321.1 427 Intron NP_001311250.1
NM_015112.2 427 Silent Mutation CGG,CGT R48R NP_055927.2
XM_005270655.3 427 Intron XP_005270712.1
XM_005270656.4 427 Intron XP_005270713.1
XM_006710477.3 427 Intron XP_006710540.1
XM_011541059.2 427 Silent Mutation CGG,CGT R48R XP_011539361.1
XM_011541061.2 427 Silent Mutation CGG,CGT R48R XP_011539363.1
XM_011541062.2 427 Silent Mutation CGG,CGT R48R XP_011539364.1
XM_011541063.1 427 Intron XP_011539365.1
XM_011541064.2 427 Intron XP_011539366.1
XM_011541065.1 427 Intron XP_011539367.1
XM_011541067.2 427 Intron XP_011539369.1
XM_011541068.1 427 Intron XP_011539370.1
XM_011541069.2 427 Silent Mutation CGG,CGT R48R XP_011539371.1
XM_017000752.1 427 Intron XP_016856241.1
XM_017000753.1 427 Intron XP_016856242.1
XM_017000754.1 427 Intron XP_016856243.1
XM_017000755.1 427 Intron XP_016856244.1

View Full Product Details