Product Details

SNP ID
rs61751018
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:50970488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCACAAAATGGATTTGGAAGGAC[C/T]GCGCTGCAGGTTGGTATTAAGAGAG
Phenotype
MIM: 603369
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDKN2C PubMed Links

Gene Details

Gene
CDKN2C
Gene Name
cyclin dependent kinase inhibitor 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001262.2 1336 Silent Mutation ACC,ACT T40T NP_001253.1
NM_078626.2 1336 Silent Mutation ACC,ACT T40T NP_523240.1

View Full Product Details