Product Details

SNP ID
rs57641470
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:53246476 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAAAAATAAATACTCTCACTTCT[C/T]TAAATTATTTATAGAACTCATCTGA
Phenotype
MIM: 602600 MIM: 602603
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100507564 PubMed Links

Gene Details

Gene
LOC100507564
Gene Name
uncharacterized LOC100507564
There are no transcripts associated with this gene.

Gene
LRP8
Gene Name
LDL receptor related protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018054.2 3228 UTR 3 NP_001018064.1
NM_004631.4 3228 UTR 3 NP_004622.2
NM_017522.4 3228 UTR 3 NP_059992.3
NM_033300.3 3228 UTR 3 NP_150643.2
XM_005271173.3 3228 UTR 3 XP_005271230.1
XM_005271174.3 3228 UTR 3 XP_005271231.1
XM_005271175.3 3228 UTR 3 XP_005271232.1
XM_006710881.3 3228 UTR 3 XP_006710944.1
XM_006710882.3 3228 UTR 3 XP_006710945.1
XM_011542094.2 3228 UTR 3 XP_011540396.1
XM_011542095.2 3228 UTR 3 XP_011540397.1
XM_011542096.2 3228 Intron XP_011540398.1
XM_017002265.1 3228 UTR 3 XP_016857754.1
XM_017002266.1 3228 Intron XP_016857755.1
XM_017002267.1 3228 UTR 3 XP_016857756.1
XM_017002268.1 3228 UTR 3 XP_016857757.1
Gene
MAGOH
Gene Name
mago homolog, exon junction complex core component
There are no transcripts associated with this gene.

View Full Product Details