Product Details

SNP ID
rs62637003
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:68431165 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAAGTTTCTTTAGTTTTGACATT[A/C]AGCTTACAGAGCTGTTTGTGAGAAA
Phenotype
MIM: 180069
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RPE65 PubMed Links

Gene Details

Gene
RPE65
Gene Name
RPE65, retinoid isomerohydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000329.2 1616 Silent Mutation CTG,CTT L450L NP_000320.1
XM_017002027.1 1616 Silent Mutation CTG,CTT L358L XP_016857516.1

View Full Product Details