Product Details
- SNP ID
-
rs61781618
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
21
- Location
-
Chr.1:92170786 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACATTCAAATTCTTATTTGAAGAGC[A/G]TAGACATGAGTTCTGCAATAAATTC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA1107
PubMed Links
Gene Details
- Gene
- KIAA1107
- Gene Name
- KIAA1107
View Full Product Details