Product Details
- SNP ID
-
hCV90452864
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:36139725 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGTGGCCCAGAATAATGACCTTGG[C/G]AATGACATCCGCAGTTTCCCGAAAG
- Phenotype
-
MIM: 610955
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TRAPPC3
PubMed Links
Gene Details
- Gene
- TRAPPC3
- Gene Name
- trafficking protein particle complex 3
There are no transcripts associated with this gene.
View Full Product Details