Product Details

SNP ID
rs61778075
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:37804330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGACGAATGATGCCAAACTTTC[C/T]GGGCACAGACAAATCAACCACAGTT
Phenotype
MIM: 600172 MIM: 612276
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf122 PubMed Links

Gene Details

Gene
C1orf122
Gene Name
chromosome 1 open reading frame 122
There are no transcripts associated with this gene.

Gene
MANEAL
Gene Name
mannosidase endo-alpha like
There are no transcripts associated with this gene.

Gene
MTF1
Gene Name
metal regulatory transcription factor 1
There are no transcripts associated with this gene.

Gene
YRDC
Gene Name
yrdC N6-threonylcarbamoyltransferase domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024640.3 752 Missense Mutation AGA,GGA R247G NP_078916.3

View Full Product Details