Product Details

SNP ID
rs56882203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:108157447 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACTGTAGTACAGTCTCAAGCTGA[A/G]TTAAATAAACATTAACTATATGATA
Phenotype
MIM: 607809 MIM: 601448
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACAT1 PubMed Links

Gene Details

Gene
ACAT1
Gene Name
acetyl-CoA acetyltransferase 1
There are no transcripts associated with this gene.

Gene
NPAT
Gene Name
nuclear protein, coactivator of histone transcription
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321307.1 6646 UTR 3 NP_001308236.1
NM_002519.2 6646 UTR 3 NP_002510.2
XM_011542854.2 6646 UTR 3 XP_011541156.1
XM_017017802.1 6646 UTR 3 XP_016873291.1
XM_017017803.1 6646 UTR 3 XP_016873292.1

View Full Product Details