Product Details

SNP ID
rs61753081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119357819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTTATACACCGTTTTCTGCCT[A/C]GGCAGCGACAGCATGTCTGAGAGAC
Phenotype
MIM: 604725
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
USP2 PubMed Links

Gene Details

Gene
USP2
Gene Name
ubiquitin specific peptidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243759.1 695 Missense Mutation NP_001230688.1
NM_004205.4 695 Missense Mutation NP_004196.4
NM_171997.2 695 Missense Mutation NP_741994.1
XM_005271721.4 695 Missense Mutation XP_005271778.1
XM_005271722.2 695 Missense Mutation XP_005271779.1
XM_017018539.1 695 Missense Mutation XP_016874028.1

View Full Product Details