Product Details

SNP ID
rs61753773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102692995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATTCCAGGAAAGTGTTTTACCA[C/T]TCTCTGCGGGAACCCTTTGTCCATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MMP27 PubMed Links

Gene Details

Gene
MMP27
Gene Name
matrix metallopeptidase 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022122.2 1263 Missense Mutation ATG,GTG M414V NP_071405.2
XM_011542948.2 1263 Missense Mutation ATG,GTG M371V XP_011541250.1
XM_011542949.2 1263 Missense Mutation ATG,GTG M360V XP_011541251.1
XM_011542950.2 1263 Missense Mutation ATG,GTG M309V XP_011541252.1
XM_017018120.1 1263 Missense Mutation ATG,GTG M413V XP_016873609.1

View Full Product Details