Product Details

SNP ID
rs61896164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55967804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTTTAGCCTTTCCTCTGGCTGA[A/T]GACAATTTCAAAATGTTGGAGATAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR10AG1 PubMed Links

Gene Details

Gene
OR10AG1
Gene Name
olfactory receptor family 10 subfamily AG member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005491.1 660 Silent Mutation TCA,TCT S220S NP_001005491.1

View Full Product Details