Product Details

SNP ID
rs58625186
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55993862 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTAGGTCTCAGGTAAGTATAGATG[C/G]AGGTGGCATAGAACAGAATTATGGC
Phenotype
MIM: 608492
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR5F1 PubMed Links

Gene Details

Gene
OR5F1
Gene Name
olfactory receptor family 5 subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003697.1 764 Missense Mutation TCC,TGC S255C NP_003688.1
Gene
OR7E5P
Gene Name
olfactory receptor family 7 subfamily E member 5 pseudogene
There are no transcripts associated with this gene.

View Full Product Details