Product Details
- SNP ID
-
rs61746318
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:56122880 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCCATGAGCAGATTGCATTTCTGT[G/T]ACTCAAACATAATTCATCACTTTTT
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR8H3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78856925] are located under a probe and SNP(s) [rs61743838] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR8H3
- Gene Name
- olfactory receptor family 8 subfamily H member 3
View Full Product Details