Product Details

SNP ID
rs61902822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58190664 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCGATCGTCGGCTCCACACCCC[A/G]ATGTACTTCTTCCTCAGCCACCTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR9Q1 PubMed Links

Gene Details

Gene
OR9Q1
Gene Name
olfactory receptor family 9 subfamily Q member 1
There are no transcripts associated with this gene.

Gene
OR9Q2
Gene Name
olfactory receptor family 9 subfamily Q member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005283.2 231 Silent Mutation CCA,CCG P58P NP_001005283.1

View Full Product Details