Product Details
- SNP ID
-
hCV90507580
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:85698075 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACTTCTCCAGTTGTAGGAAGCTTT[C/T]TACCTACAATAGAAAAAGAAGAGAA
- Phenotype
-
MIM: 612880
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SYTL2
PubMed Links
Gene Details
- Gene
- SYTL2
- Gene Name
- synaptotagmin like 2
There are no transcripts associated with this gene.
View Full Product Details