Product Details

SNP ID
rs56069251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:44915989 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATGGTCATTGTCTCGCTCCAGCC[C/T]CACAAATGCCTGAGACGCTAGAGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM72 PubMed Links

Gene Details

Gene
TMEM72
Gene Name
transmembrane protein 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123376.2 Intron NP_001116848.1
XM_011540069.2 Intron XP_011538371.1
XM_011540070.2 Intron XP_011538372.1
XM_011540071.2 Intron XP_011538373.1
Gene
TMEM72-AS1
Gene Name
TMEM72 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details