Product Details

SNP ID
rs55774248
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134196654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAATCTGTCAATCATTTTAAAGAG[A/T]CCATATAATATACACATAGCAATAT
Phenotype
MIM: 176915
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PPP2CA PubMed Links

Gene Details

Gene
PPP2CA
Gene Name
protein phosphatase 2 catalytic subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002715.2 2446 UTR 3 NP_002706.1

View Full Product Details