Product Details

SNP ID
rs62375426
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:137618758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCACCCCCACAGAAAGCAGACT[A/C]CAAAAAAAAAAAAAAAGACACAATC
Phenotype
MIM: 605775
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
KLHL3 PubMed Links
Additional Information
For this assay, SNP(s) [rs58917494] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
KLHL3
Gene Name
kelch like family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257194.1 5548 UTR 3 NP_001244123.1
NM_001257195.1 5548 UTR 3 NP_001244124.1
NM_017415.2 5548 UTR 3 NP_059111.2

View Full Product Details