Product Details

SNP ID
rs62381765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138188561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATCCATTGCTCTGACTTCTTCCT[A/G]GGGTACTCAGTGCCCTACTGCCAGC
Phenotype
MIM: 602848 MIM: 603462 MIM: 605664
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRD8 PubMed Links

Gene Details

Gene
BRD8
Gene Name
bromodomain containing 8
There are no transcripts associated with this gene.

Gene
CDC23
Gene Name
cell division cycle 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004661.3 2242 UTR 3 NP_004652.2
Gene
KIF20A
Gene Name
kinesin family member 20A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005733.2 2242 Intron NP_005724.1

View Full Product Details