Product Details

SNP ID
rs59473271
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:139372530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTTCATTATCTTGTTTCTTGAA[A/G]ACATACCCCTGGGAGATGTATAGAC
Phenotype
MIM: 605604 MIM: 603790
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PAIP2 PubMed Links

Gene Details

Gene
PAIP2
Gene Name
poly(A) binding protein interacting protein 2
There are no transcripts associated with this gene.

Gene
SLC23A1
Gene Name
solute carrier family 23 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005847.4 Intron NP_005838.3
NM_152685.3 Intron NP_689898.2
XM_005272148.3 Intron XP_005272205.3
XM_005272149.3 Intron XP_005272206.3
XM_006714741.2 Intron XP_006714804.2
XM_011543765.2 Intron XP_011542067.1
XM_011543766.1 Intron XP_011542068.1
XM_011543767.1 Intron XP_011542069.1
XM_011543768.1 Intron XP_011542070.1
XM_011543769.1 Intron XP_011542071.1

View Full Product Details