Product Details

SNP ID
rs57802226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:130719901 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAACTTTTTGCCAGAATCATTC[A/G]GGGTCAAAGGGCTAGTCATATTTAC
Phenotype
MIM: 189980
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABL1 PubMed Links

Gene Details

Gene
ABL1
Gene Name
ABL proto-oncogene 1, non-receptor tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005157.5 Intron NP_005148.2
NM_007313.2 Intron NP_009297.2

View Full Product Details