Product Details

SNP ID
rs56879537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1364545 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTCAACTCCTAGGGACAGTCCCA[A/G]CCCTGAGCAGAAGGGTCAGGTCATC
Phenotype
MIM: 607838
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNPTG PubMed Links

Gene Details

Gene
GNPTG
Gene Name
N-acetylglucosamine-1-phosphate transferase gamma subunit
There are no transcripts associated with this gene.

Gene
UNKL
Gene Name
unkempt family like zinc finger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037125.3 3955 Intron NP_001032202.1
NM_001193388.3 3955 UTR 3 NP_001180317.2
NM_001193389.1 3955 UTR 3 NP_001180318.1
NM_001276414.1 3955 UTR 3 NP_001263343.1
XM_005255505.1 3955 UTR 3 XP_005255562.1
XM_011522610.1 3955 UTR 3 XP_011520912.1
XM_011522611.1 3955 UTR 3 XP_011520913.1
XM_011522612.1 3955 UTR 3 XP_011520914.1
XM_011522613.1 3955 Intron XP_011520915.1
XM_017023567.1 3955 Intron XP_016879056.1
XM_017023568.1 3955 UTR 3 XP_016879057.1
XM_017023569.1 3955 UTR 3 XP_016879058.1

View Full Product Details