Product Details
- SNP ID
-
hCV90591004
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:2027216 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAACGTGGAGGGCGAGACGCACCAC[C/T]AGGTGGGGGCCAGCGCTCGCCCCCG
- Phenotype
-
MIM: 607997
MIM: 606553
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NPW
PubMed Links
Gene Details
- Gene
- NPW
- Gene Name
- neuropeptide W
There are no transcripts associated with this gene.
- Gene
- SLC9A3R2
- Gene Name
- SLC9A3 regulator 2
There are no transcripts associated with this gene.
View Full Product Details