Product Details

SNP ID
rs60782450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3541410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAAACGGATGCTCCTCACGGGCT[G/T]GAGGTGGCAGCTCATCTCAGAGCAT
Phenotype
MIM: 616787 MIM: 615648
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLUAP1 PubMed Links

Gene Details

Gene
CLUAP1
Gene Name
clusterin associated protein 1
There are no transcripts associated with this gene.

Gene
LOC101929732
Gene Name
uncharacterized LOC101929732
There are no transcripts associated with this gene.

Gene
NLRC3
Gene Name
NLR family CARD domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178844.3 3962 UTR 3 NP_849172.2
XM_017023027.1 3962 UTR 3 XP_016878516.1
XM_017023028.1 3962 UTR 3 XP_016878517.1
XM_017023029.1 3962 UTR 3 XP_016878518.1
XM_017023030.1 3962 UTR 3 XP_016878519.1
XM_017023031.1 3962 UTR 3 XP_016878520.1
XM_017023032.1 3962 UTR 3 XP_016878521.1
XM_017023033.1 3962 UTR 3 XP_016878522.1
XM_017023034.1 3962 UTR 3 XP_016878523.1
XM_017023035.1 3962 UTR 3 XP_016878524.1
XM_017023036.1 3962 UTR 3 XP_016878525.1
XM_017023037.1 3962 UTR 3 XP_016878526.1
XM_017023038.1 3962 UTR 3 XP_016878527.1
XM_017023039.1 3962 Intron XP_016878528.1

View Full Product Details