Product Details
- SNP ID
-
rs62323388
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:70594339 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTTGAACACTGAACTACTCCAGC[A/C]AACAGCATGCAATTCTGTTATCACA
- Phenotype
-
MIM: 601259
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
AMBN
PubMed Links
Gene Details
- Gene
- AMBN
- Gene Name
- ameloblastin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016519.5 |
|
Intron |
|
|
NP_057603.1 |
View Full Product Details