Product Details

SNP ID
rs58371816
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:69220498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTACACATTTTGCACTCACACTG[C/T]GCTAACAGAGTTTCTTCATGATTGT
Phenotype
MIM: 603064
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105377267 PubMed Links

Gene Details

Gene
LOC105377267
Gene Name
uncharacterized LOC105377267
There are no transcripts associated with this gene.

Gene
UGT2B11
Gene Name
UDP glucuronosyltransferase family 2 member B11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011531550.2 Intron XP_011529852.1
XM_017007660.1 Intron XP_016863149.1
XM_017007661.1 Intron XP_016863150.1

View Full Product Details