Product Details

SNP ID
rs58573983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:11422445 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCAAGGGCTTATCTGGAAAGAGG[C/T]TGGCAGGAAAGGAATGCGTCTTCAG
Phenotype
MIM: 610085
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM167A PubMed Links

Gene Details

Gene
FAM167A
Gene Name
family with sequence similarity 167 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_053279.2 3465 UTR 3 NP_444509.2
XM_005272398.4 3465 UTR 3 XP_005272455.1
XM_011543837.1 3465 UTR 3 XP_011542139.1
XM_011543838.2 3465 UTR 3 XP_011542140.1
XM_011543840.2 3465 UTR 3 XP_011542142.1
Gene
FAM167A-AS1
Gene Name
FAM167A antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details