Product Details

SNP ID
rs72634820
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:1665581 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACACAGTCAGCTACTGGTCTGGT[C/G]TCTACTCCAGGAAGCTGATACCTGG
Phenotype
MIM: 176873
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDK11B PubMed Links

Gene Details

Gene
CDK11B
Gene Name
cyclin dependent kinase 11B
There are no transcripts associated with this gene.

Gene
SLC35E2B
Gene Name
solute carrier family 35 member E2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001110781.2 2340 UTR 3 NP_001104251.1
NM_001290264.1 2340 UTR 3 NP_001277193.1
XM_011542072.1 2340 UTR 3 XP_011540374.1

View Full Product Details