Product Details

SNP ID
rs74125561
Assay Type
Functionally tested
NCBI dbSNP Submissions
27
Location
Chr.1:160279572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTCACATGATCAGACACTGTTC[A/G]CCACTGGCACCTGGTGGGCCCGAAA
Phenotype
MIM: 601924 MIM: 600279
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COPA PubMed Links

Gene Details

Gene
COPA
Gene Name
coatomer protein complex subunit alpha
There are no transcripts associated with this gene.

Gene
PEX19
Gene Name
peroxisomal biogenesis factor 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193644.1 906 Missense Mutation GCG,GTG A276V NP_001180573.1
NM_002857.3 906 Silent Mutation GGC,GGT G293G NP_002848.1

View Full Product Details