Product Details

SNP ID
rs71632311
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:167430755 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGTTTTCCATCCATGGCCTGTGC[C/T]CTGTAATGACGCAGCAGTATCCTAG
Phenotype
MIM: 186780 MIM: 164175
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD247 PubMed Links

Gene Details

Gene
CD247
Gene Name
CD247 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000734.3 1640 UTR 3 NP_000725.1
NM_198053.2 1640 UTR 3 NP_932170.1
XM_011510144.2 1640 UTR 3 XP_011508446.1
XM_011510145.1 1640 UTR 3 XP_011508447.1
XM_017002800.1 1640 UTR 3 XP_016858289.1
XM_017002801.1 1640 UTR 3 XP_016858290.1
Gene
POU2F1
Gene Name
POU class 2 homeobox 1
There are no transcripts associated with this gene.

View Full Product Details