Product Details
- SNP ID
-
rs74121074
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:168229959 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGGCTCATCTGTCAGATCTTAAC[A/G]CTGGGCCCCTGCTGTCTGTTATCAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SFT2D2
PubMed Links
Gene Details
- Gene
- SFT2D2
- Gene Name
- SFT2 domain containing 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_199344.2 |
|
Intron |
|
|
NP_955376.1 |
View Full Product Details