Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001135821.1 | 98 | Intron | NP_001129293.1 | ||
NM_001135822.1 | 98 | Intron | NP_001129294.1 | ||
NM_001242824.1 | 98 | Intron | NP_001229753.1 | ||
NM_001242825.1 | 98 | Intron | NP_001229754.1 | ||
NM_002004.3 | 98 | Intron | NP_001995.1 | ||
XM_005244962.1 | 98 | Intron | XP_005245019.1 | ||
XM_005244963.1 | 98 | Intron | XP_005245020.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001105203.1 | 98 | Intron | NP_001098673.1 | ||
NM_001105204.1 | 98 | Intron | NP_001098674.1 | ||
NM_001105205.1 | 98 | Intron | NP_001098675.1 | ||
NM_001278227.1 | 98 | Intron | NP_001265156.1 | ||
NM_001278228.1 | 98 | Intron | NP_001265157.1 | ||
NM_001278229.1 | 98 | Intron | NP_001265158.1 | ||
NM_001278230.1 | 98 | Intron | NP_001265159.1 | ||
NM_014328.4 | 98 | Intron | NP_055143.2 | ||
XM_006711254.1 | 98 | Intron | XP_006711317.1 | ||
XM_006711256.1 | 98 | Intron | XP_006711319.1 | ||
XM_006711257.1 | 98 | Intron | XP_006711320.1 | ||
XM_017000891.1 | 98 | UTR 5 | XP_016856380.1 | ||
XM_017000892.1 | 98 | Intron | XP_016856381.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001039517.1 | 98 | Nonsense Mutation | TAA,TCA | *6S | NP_001034606.1 |