Product Details

SNP ID
rs67195253
Assay Type
Functionally tested
NCBI dbSNP Submissions
31
Location
Chr.1:156827023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAAAAGTAAATAAAAACTCAGCT[C/T]CTTGTTTTGGAGCTCACTTCTGGGG
Phenotype
MIM: 191315
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NTRK1 PubMed Links

Gene Details

Gene
NTRK1
Gene Name
neurotrophic receptor tyrosine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007792.1 Intron NP_001007793.1
NM_001012331.1 Intron NP_001012331.1
NM_002529.3 Intron NP_002520.2

View Full Product Details