Product Details

SNP ID
rs73013119
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:150300021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAAAAAACATACCATGAGATTTC[A/G]CCAGGATTGAAAATTATTATAATAG
Phenotype
MIM: 611984
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPS21 PubMed Links

Gene Details

Gene
MRPS21
Gene Name
mitochondrial ribosomal protein S21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018997.3 Intron NP_061870.1
NM_031901.5 Intron NP_114107.1

View Full Product Details