Product Details

SNP ID
rs73019729
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:158420329 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGCTTGAGGACAGGAGCAATGT[C/T]ACAGAAGAAGTGATGTAGTTGATTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10K2 PubMed Links

Gene Details

Gene
OR10K2
Gene Name
olfactory receptor family 10 subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004476.1 538 Missense Mutation AAC,GAC N180D NP_001004476.1

View Full Product Details