Product Details
- SNP ID
-
rs71642227
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
24
- Location
-
Chr.1:28262272 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTCACCACACACTCCCTTTCAGGA[G/T]CTCATTGCCTTCAGATGTGTGTATC
- Phenotype
-
MIM: 607767
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SESN2
PubMed Links
Gene Details
- Gene
- SESN2
- Gene Name
- sestrin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_031459.4 |
|
Intron |
|
|
NP_113647.1 |
View Full Product Details