Product Details

SNP ID
rs72889606
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:28819637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCTCATTTGCAAATGGGGACAA[A/G]AAGGCTCAGAGGGAGCGAGTGCCTC
Phenotype
MIM: 165195
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OPRD1 PubMed Links

Gene Details

Gene
OPRD1
Gene Name
opioid receptor delta 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000911.3 Intron NP_000902.3

View Full Product Details