Product Details

SNP ID
rs74062789
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:23807386 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAACAGACTCACCTGGCTTAGACA[C/T]GAACCTCTAGGGGATAATTGAGAGG
Phenotype
MIM: 606953 MIM: 613898
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GALE PubMed Links

Gene Details

Gene
GALE
Gene Name
UDP-galactose-4-epimerase
There are no transcripts associated with this gene.

Gene
HMGCL
Gene Name
3-hydroxymethyl-3-methylglutaryl-CoA lyase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000191.2 Intron NP_000182.2
NM_001166059.1 Intron NP_001159531.1

View Full Product Details