Product Details

SNP ID
rs71642416
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:247491749 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTGCCCTGGGGGTGGCTCTCCT[C/T]CTGGTGCCGCTCTCCCTCATCCTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2W5 PubMed Links

Gene Details

Gene
OR2W5
Gene Name
olfactory receptor family 2 subfamily W member 5 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004698.2 682 Missense Mutation CCT,TCT P208S NP_001004698.1

View Full Product Details